Anti-ArhGEF6 Rabbit Monoclonal Antibody [clone: C23D2]
CSTA4573S
- Antibody type:Primary
- Antigen symbol:ArhGEF6
- Clonality:Monoclonal
- Clone:C23D2
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Sequence:CGYKERMSYILKESSKSPKT
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: ArhGEF6
Clonality: Monoclonal
Clone: C23D2
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse