Anti-Helios Rabbit Monoclonal Antibody [clone: D8W4X]
Type: Primary
Antigen: Helios
Clonality: Monoclonal
Clone: D8W4X
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- CSTA42427S
- CSTA42427T
- Antigen symbol:
- Helios
- Helios
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- D8W4X
- D8W4X
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- ChIP:
- Yes
- Yes
- Sequence:
- CDRKPLSREDEIRGHDEGSSLE
- CDRKPLSREDEIRGHDEGSSLE
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Ambient
- Ambient
- Tested applications:
- CUT&RUN
- CUT&RUN
Frequently Bought Together