Anti-APOBEC3B Rabbit Monoclonal Antibody [clone: E9A2G]
Catalog # CSTA41494S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:APOBEC3B
- Clonality:Monoclonal
- Clone:E9A2G
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Sequence:CNFNNDPLVLRRRQTYL
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: APOBEC3B
Clonality: Monoclonal
Clone: E9A2G
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human