Anti-p27 Kip1 Rabbit Monoclonal Antibody [clone: D69C12]
Supplier: Cell Signaling
Certificates
About this item
Type: Primary
Antigen: p27 Kip1
Clonality: Monoclonal
Clone: D69C12
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Monkey
Specifications
- Catalog No:
- CSTA3686S
- CSTA3686T
- Antigen symbol:
- p27 Kip1
- p27 Kip1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- D69C12
- D69C12
- Reactivity:
- Monkey, Human, Rat
- Monkey, Human, Rat
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Sequence:
- PSLERMDARQAEHPKPSAC
- PSLERMDARQAEHPKPSAC
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together