Anti-PSD-93 Rabbit Monoclonal Antibody [clone: D4Z4D]
CSTA19046S
- Antibody type:Primary
- Antigen symbol:PSD-93
- Clonality:Monoclonal
- Clone:D4Z4D
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Sequence:CNNGTLEYKTSLPPISPGRYSP
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: PSD-93
Clonality: Monoclonal
Clone: D4Z4D
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat