Anti-HSL Rabbit Monoclonal Antibody [clone: D6W5S]
Supplier: Cell Signaling
Certificates
About this item
Type: Primary
Antigen: HSL
Clonality: Monoclonal
Clone: D6W5S
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- CSTA18381S
- CSTA18381T
- Antigen symbol:
- HSL
- HSL
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- D6W5S
- D6W5S
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Sequence:
- CGVRAAFPEGFHPRRSSQG
- CGVRAAFPEGFHPRRSSQG
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together