Anti-PDCD8 Rabbit Monoclonal Antibody [clone: D39D2]
Catalog # CSTA62147SF
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:PDCD8
- Clonality:Monoclonal
- Clone:D39D2
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:CDNPKSATEQSGTGIRSES
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:100 µG
Specifications
About this item
- BSA and Azide Free
Type: Primary
Antigen: PDCD8
Clonality: Monoclonal
Clone: D39D2
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey