Anti-G4S Linker Rabbit Monoclonal Antibody [clone: E7O2V] (PE)
Supplier: Cell Signaling
Certificates
About this item
Type: Primary
Antigen: G4S Linker
Clonality: Monoclonal
Clone: E7O2V
Conjugation: PE
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Any species
Specifications
- Catalog No:
- CSTA38907L
- CSTA38907S
- Antigen symbol:
- G4S Linker
- G4S Linker
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- E7O2V
- E7O2V
- Reactivity:
- Multiple
- Multiple
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Flow cytometry:
- Yes
- Yes
- Sequence:
- CGGGGSGGGGSGGGGS
- CGGGGSGGGGSGGGGS
- Storage temperature:
- +4 °C
- +4 °C
- Shipping temperature:
- Cold
- Cold
Specifications
Frequently Bought Together