Anti-Whitlow Linker Rabbit - new Monoclonal Antibody [clone: E3U7Q] (PE)
Supplier: Cell Signaling
Certificates
About this item
Type: Primary
Antigen: Whitlow Linker
Clonality: Monoclonal
Clone: E3U7Q
Conjugation: PE
Epitope:
Host: Rabbit - new
Isotype: IgG
Reactivity: Any species
Specifications
- Catalog No:
- CSTA62405L
- CSTA62405S
- Antigen symbol:
- Whitlow Linker
- Whitlow Linker
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- E3U7Q
- E3U7Q
- Reactivity:
- Multiple
- Multiple
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Flow cytometry:
- Yes
- Yes
- Sequence:
- CGSTSGSGKPGSGEGSTKG
- CGSTSGSGKPGSGEGSTKG
- Storage temperature:
- +4 °C
- +4 °C
- Shipping temperature:
- Cold
- Cold
Specifications
Frequently Bought Together