Anti-NALP1 Rabbit Monoclonal Antibody [clone: E3F2U]
CSTA56719S
- Antibody type:Primary
- Antigen symbol:NALP1
- Clonality:Monoclonal
- Clone:E3F2U
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Sequence:Q9C000 A1365-S1473
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: NALP1
Clonality: Monoclonal
Clone: E3F2U
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human