Anti-Flag-Tag Rabbit Monoclonal Antibody [clone: D6W5B] (Magnetic Bead)
CSTA82103S
- Antibody type:Primary
- Antigen symbol:Flag-Tag
- Clonality:Monoclonal
- Clone:D6W5B
- Conjugation:Magnetic beads
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Multiple
- Sequence:CXXXDYKDDDDK
- Storage temperature:+4 °C
- Shipping temperature:Cold
- Pk:400 µl
Type: Primary
Antigen: Flag-Tag
Clonality: Monoclonal
Clone: D6W5B
Conjugation: Magnetic Bead
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Any species