Anti-NRF2 Rabbit Monoclonal Antibody [clone: E3J1V]
Catalog # CSTA33649S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:NRF2
- Clonality:Monoclonal
- Clone:E3J1V
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat
- Western blot:Yes
- Sequence:CPGHRKTPFTKDKHSSRLE
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: NRF2
Clonality: Monoclonal
Clone: E3J1V
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Monkey