Anti-coronin 1A Rabbit Monoclonal Antibody [clone: D6K5B] (PE)
CSTA98455S
- Antibody type:Primary
- Antigen symbol:coronin 1A
- Clonality:Monoclonal
- Clone:D6K5B
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Sequence:CRDKRVRIIEPRKGTVVAEKDRPHEGT
- Storage temperature:+4 °C
- Shipping temperature:Cold
- Pk:100 µl
Type: Primary
Antigen: coronin 1A
Clonality: Monoclonal
Clone: D6K5B
Conjugation: PE
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse