Anti-IRF-1 Rabbit Monoclonal Antibody [clone: D5E4]
Supplier: Cell Signaling
Certificates
About this item
Type: Primary
Antigen: IRF-1
Clonality: Monoclonal
Clone: D5E4
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- CSTA8478T
- CSTA8478S
- Antigen symbol:
- IRF-1
- IRF-1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- D5E4
- D5E4
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Sequence:
- CDGKGYLLNEPGVQPTSVYGDFS
- CDGKGYLLNEPGVQPTSVYGDFS
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Ambient
- Ambient
- Tested applications:
- CUT&RUN
- CUT&RUN
Specifications
Frequently Bought Together