Anti-HOP Rabbit Monoclonal Antibody [clone: D10E2]
Catalog # CSTA5670S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:HOP
- Clonality:Monoclonal
- Clone:D10E2
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Bovine
- Western blot:Yes
- Sequence:CKHYTEAIKRNPKDAKLYSNR
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: HOP
Clonality: Monoclonal
Clone: D10E2
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Bovine