Anti-PD-L1 Rabbit Monoclonal Antibody [clone: E1L3N] (HRP)
Catalog # CSTA51296S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:PD-L1
- Clonality:Monoclonal
- Clone:E1L3N
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Sequence:CGIQDTNSKKQSDTHLEET
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: PD-L1
Clonality: Monoclonal
Clone: E1L3N
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human