Anti-NAD-dependent malic enzyme, mitochondrial Rabbit Monoclonal Antibody [clone: E1N3F]
Supplier: Cell Signaling
Certificates
About this item
Type: Primary
Antigen: NAD-dependent malic enzyme, mitochondrial
Clonality: Monoclonal
Clone: E1N3F
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey
Specifications
- Catalog No:
- CSTA35939T
- CSTA35939S
- Antigen symbol:
- NAD-dependent malic enzyme, mitochondrial
- NAD-dependent malic enzyme, mitochondrial
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- E1N3F
- E1N3F
- Reactivity:
- Monkey, Human
- Monkey, Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Sequence:
- CRSEYDSLLPDVYEWPESASSP
- CRSEYDSLLPDVYEWPESASSP
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together