Anti-MCP-1 Rabbit Polyclonal Antibody
Catalog # CSTA2029S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:MCP-1
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Sequence:CADPKKEWVQTYIKNLDRNQMRSEPT
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: MCP-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse