Anti-B-Raf Rabbit Monoclonal Antibody [clone: D9T6S]
Catalog # CSTA14814S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:B-Raf
- Clonality:Monoclonal
- Clone:D9T6S
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:hb-Raf(M 1 S 180)
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: B-Raf
Clonality: Monoclonal
Clone: D9T6S
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey