Anti-Syk Rabbit Monoclonal Antibody [clone: D3Z1E] (Pacific Blue)
CSTA13722S
- Antibody type:Primary
- Antigen symbol:Syk
- Clonality:Monoclonal
- Clone:D3Z1E
- Conjugation:Pacific Blue™
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Sequence:CGPLNKYLQQNRHVKDKNIIE
- Storage temperature:+4 °C
- Shipping temperature:Cold
- Pk:100 µl
Type: Primary
Antigen: Syk
Clonality: Monoclonal
Clone: D3Z1E
Conjugation: Pacific Blue
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat