Anti-HSP27 Mouse Monoclonal Antibody [clone: G31]
CSTA47289SF
- Antibody type:Primary
- Antigen symbol:HSP27
- Clonality:Monoclonal
- Clone:G31
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Monkey,Human
- Western blot:Yes
- Sequence:UNKNOWN
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Size:100 µg
- Pk:100 µG
Type: Primary Antibodies
Antigen: HSP27
Clonality: Monoclonal
Clone: G31
Conjugation:
Epitope:
Hoat: Mouse
Isotype: IgG1
Reactivity: Human,Monkey