Anti-CRACC/SLAMF7 Rabbit Monoclonal Antibody [clone: E5C4M]
CSTA66110SF
- Antibody type:Primary
- Antigen symbol:CRACC/SLAMF7
- Clonality:Monoclonal
- Clone:E5C4M
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Sequence:Q9NQ25 A2-M226
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Size:100 µg
- Pk:100 µG
Type: Primary Antibodies
Antigen: CRACC/SLAMF7
Clonality: Monoclonal
Clone: E5C4M
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human