Anti-MAGE-A10 Rabbit Monoclonal Antibody [clone: E9V2Q]
CSTA81740S
- Antibody type:Primary
- Antigen symbol:MAGE-A10
- Clonality:Monoclonal
- Clone:E9V2Q
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Sequence:CFLAKVNGSDPRSFPLWYEE
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Size:100 µl
- Pk:100 µl
Type: Primary Antibodies
Antigen: MAGE-A10
Clonality: Monoclonal
Clone: E9V2Q
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human