Anti-ATG16L1 Rabbit Monoclonal Antibody [clone: D6D5]
Supplier: Cell Signaling
Certificates
About this item
Type: Primary Antibodies
Antigen: ATG16L1
Clonality: Monoclonal
Clone: D6D5
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat
Specifications
- Catalog No:
- CSTA8089S
- CSTA8089T
- Antigen symbol:
- ATG16L1
- ATG16L1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- D6D5
- D6D5
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Sequence:
- CNKLLEKSDLHSVLAQKLQAEKHD
- CNKLLEKSDLHSVLAQKLQAEKHD
- Storage temperature:
- −20 °C
- −20 °C
- Shipping temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together