Anti-CDT1 Rabbit Monoclonal Antibody [clone: D10F11]
CSTA8064S
- Antibody type:Primary
- Antigen symbol:CDT1
- Clonality:Monoclonal
- Clone:D10F11
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human
- Western blot:Yes
- Sequence:SRKRARPPAAPGRDQARPPAC
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Size:100 µl
- Pk:100 µl
Type: Primary Antibodies
Antigen: CDT1
Clonality: Monoclonal
Clone: D10F11
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human,Monkey