Anti-tubulin alpha (Lys40) acetylate Rabbit Monoclonal Antibody [clone: 'D20G3]
Type: Primary Antibodies
Antigen: tubulin alpha (Lys40) acetylate
Clonality: Monoclonal
Clone: 'D20G3
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat,Monkey,Zebrafish
- Catalog No:
- CSTA5335S
- CSTA5335T
- Antigen symbol:
- tubulin alpha (Lys40) acetylate
- tubulin alpha (Lys40) acetylate
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- D20G3
- D20G3
- Reactivity:
- Zebrafish, Monkey, Human, Rat, Mouse
- Zebrafish, Monkey, Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Sequence:
- CQPDGQMPSD(K-Ac)TIGG
- CQPDGQMPSD(K-Ac)TIGG
- Storage temperature:
- −20 °C
- −20 °C
- Shipping temperature:
- Ambient
- Ambient
Frequently Bought Together