Anti-4E-BP2 (Thr46) non-phosphate Rabbit Monoclonal Antibody [clone: '87D12]
Supplier: Cell Signaling
Certificates
About this item
Type: Primary Antibodies
Antigen: 4E-BP2 (Thr46) non-phosphate
Clonality: Monoclonal
Clone: '87D12
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat,Monkey
Specifications
- Catalog No:
- CSTA4923S
- CSTA4923T
- Antigen symbol:
- 4E-BP2 (Thr46) non-phosphate
- 4E-BP2 (Thr46) non-phosphate
- Antigen synonyms:
- 4E-BP2 (Thr46) non-phosphate|4E-BP1 (Thr46) non-phosphate|4E-BP3 (Thr32) non-phosphate
- 4E-BP2 (Thr46) non-phosphate|4E-BP1 (Thr46) non-phosphate|4E-BP3 (Thr32) non-phosphate
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 87D12
- 87D12
- Reactivity:
- Monkey, Human, Rat, Mouse
- Monkey, Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Sequence:
- CDYSTTPGGTLFSTTPGGTRIIYDRK
- CDYSTTPGGTLFSTTPGGTRIIYDRK
- Storage temperature:
- −20 °C
- −20 °C
- Shipping temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together