Anti-HIP Rabbit Polyclonal Antibody
CSTA2723S
- Antibody type:Primary
- Antigen symbol:HIP
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:CDRAIEINPDSAQPYKWRGKAHR
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Size:100 µl
- Pk:100 µl
Type: Primary Antibodies
Antigen: HIP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Hoat: Rabbit
Isotype:
Reactivity: Human,Mouse,Rat,Monkey