Anti-OLFM4 Rabbit Monoclonal Antibody [clone: D1E4M]
Supplier: Cell Signaling
Certificates
About this item
Type: Primary Antibodies
Antigen: OLFM4
Clonality: Monoclonal
Clone: D1E4M
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- CSTA14369T
- CSTA14369S
- Antigen symbol:
- OLFM4
- OLFM4
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- D1E4M
- D1E4M
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Sequence:
- CSVSLPDTTFPVDRVER
- CSVSLPDTTFPVDRVER
- Storage temperature:
- −20 °C
- −20 °C
- Shipping temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together