Anti-GPR50 Rabbit Monoclonal Antibody [clone: D1D6I]
Catalog # CSTA14032S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:GPR50
- Clonality:Monoclonal
- Clone:D1D6I
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Sequence:CSHSKAASGHLKPVSGHSKPASGHPK
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary Antibodies
Antigen: GPR50
Clonality: Monoclonal
Clone: D1D6I
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human