Anti-p130Cas Rabbit Monoclonal Antibody [clone: E1L9G]
Catalog # CSTA13383S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:p130Cas
- Clonality:Monoclonal
- Clone:E1L9G
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat
- Western blot:Yes
- Sequence:CYQPQPDSVYLVPTPSKAQQG
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary Antibody Conjugates
Antigen: p130Cas
Clonality: Monoclonal
Clone: E1L9G
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human,Rat,Monkey