Anti-CP110 Rabbit Polyclonal Antibody
Catalog # CSTA12140S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:CP110
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Monkey,Human
- Western blot:Yes
- Sequence:CDNSFEKVKRRLDLDIDGLQKE
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary Antibodies
Antigen: CP110
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Hoat: Rabbit
Isotype:
Reactivity: Human,Monkey