Anti-CPSF4 Rabbit Monoclonal Antibody [clone: E2L2Z]
Catalog # CSTA70997S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:CPSF4
- Clonality:Monoclonal
- Clone:E2L2Z
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:CTKGHLAFLSGQ
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: CPSF4
Clonality: Monoclonal
Clone: E2L2Z
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey