Anti-HPK1 Rabbit Monoclonal Antibody [clone: E1C3L] (Alexa Fluor 647)
Catalog # CSTA93808S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:HPK1
- Clonality:Monoclonal
- Clone:E1C3L
- Conjugation:Alexa Fluor® 647
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Sequence:VDPDIFNRDPRDHYDLLQRC
- Storage temperature:4 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: HPK1
Clonality: Monoclonal
Clone: E1C3L
Conjugation: Alexa Fluor 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human