Anti-GPR37 Rabbit Monoclonal Antibody [clone: D4C8H]
Catalog # CSTA64407S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:GPR37
- Clonality:Monoclonal
- Clone:D4C8H
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Sequence:CNDNEYTTELELSPFSTIRR
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: GPR37
Clonality: Monoclonal
Clone: D4C8H
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat