Anti-Hemoglobin subunit gamma Rabbit Monoclonal Antibody [clone: D4K7X] (Alexa Fluor 488)
Catalog # CSTA51918S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Hemoglobin subunit gamma
- Clonality:Monoclonal
- Clone:D4K7X
- Conjugation:Alexa Fluor® 488
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Sequence:CTSLWGKVNVEDAGGETLG
- Storage temperature:4 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Hemoglobin subunit gamma
Clonality: Monoclonal
Clone: D4K7X
Conjugation: Alexa Fluor 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human