Anti-ULK1 Rabbit Polyclonal Antibody
Supplier: Cell Signaling
Certificates
About this item
Type: Primary
Antigen: ULK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- CSTA4776S
- CSTA4776T
- Antigen symbol:
- ULK1
- ULK1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- Sequence:
- LKAAFGTQAPDPGSTESLC
- LKAAFGTQAPDPGSTESLC
- Storage temperature:
- −20 °C
- −20 °C
- Shipping temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together