Anti-Notch1 (Val1754,N-term) cleaved Rabbit Monoclonal Antibody [clone: D3B8]
Supplier: Cell Signaling
Certificates
About this item
Type: Primary
Antigen: Notch1 (Val1754,N-term) cleaved
Clonality: Monoclonal
Clone: D3B8
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- CSTA4147S
- CSTA4147T
- Antigen symbol:
- Notch1 (Val1754,N-term) cleaved
- Notch1 (Val1754,N-term) cleaved
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- D3B8
- D3B8
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ChIP:
- Yes
- Yes
- Sequence:
- VLLSRKRRRQHGQLC
- VLLSRKRRRQHGQLC
- Storage temperature:
- −20 °C
- −20 °C
- Shipping temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together