Anti-TIF1b Rabbit Monoclonal Antibody [clone: C42G12]
Supplier: Cell Signaling
Certificates
About this item
Type: Primary
Antigen: TIF1b
Clonality: Monoclonal
Clone: C42G12
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey
Specifications
- Catalog No:
- CSTA4124S
- CSTA4124T
- Antigen symbol:
- TIF1b
- TIF1b
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- C42G12
- C42G12
- Reactivity:
- Monkey, Human, Rat, Mouse
- Monkey, Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Sequence:
- CAEGPGAEGPRLASPSGSTSSG
- CAEGPGAEGPRLASPSGSTSSG
- Storage temperature:
- −20 °C
- −20 °C
- Shipping temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together