Anti-IRP1 Rabbit Monoclonal Antibody [clone: D6S4J]
CSTA20272S
- Antibody type:Primary
- Antigen symbol:IRP1
- Clonality:Monoclonal
- Clone:D6S4J
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Sequence:CANIRLLNRFLNKQAPQTIHLPSGEILD
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: IRP1
Clonality: Monoclonal
Clone: D6S4J
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human