Anti-MIOS Rabbit Monoclonal Antibody [clone: D12C6]
CSTA13557S
- Antibody type:Primary
- Antigen symbol:MIOS
- Clonality:Monoclonal
- Clone:D12C6
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Sequence:CHKRAEFDIHRSKLDPSSKPLA
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: MIOS
Clonality: Monoclonal
Clone: D12C6
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat