Anti-HES1 Rabbit Monoclonal Antibody [clone: D6P2U]
Catalog # CSTA33699SF
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:HES1
- Clonality:Monoclonal
- Clone:D6P2U
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:hHES(M1 to N277)
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Pk:100 µG
Specifications
About this item
- BSA and azide free
Type: Primary
Antigen: HES1
Clonality: Monoclonal
Clone: D6P2U
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey