Anti-gasdermin D (Asp275) cleaved Rabbit Monoclonal Antibody [clone: E7H9G]
CSTA37349SF
- Antibody type:Primary
- Antigen symbol:gasdermin D (Asp275) cleaved
- Clonality:Monoclonal
- Clone:E7H9G
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Sequence:CLHNFLTD
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Pk:100 µG
- BSA and azide free
Type: Primary
Antigen: gasdermin D (Asp275) cleaved
Clonality: Monoclonal
Clone: E7H9G
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human