Anti-PSMB9 Rabbit Monoclonal Antibody [clone: E7J1L]
Catalog # CSTA87667S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:PSMB9
- Clonality:Monoclonal
- Clone:E7J1L
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Sequence:CRVILG(N/D)ELPKFYDE
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: PSMB9
Clonality: Monoclonal
Clone: E7J1L
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat