Anti-ETV5/ERM Rabbit Monoclonal Antibody [clone: E5G9V]
Catalog # CSTA16274S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:ETV5/ERM
- Clonality:Monoclonal
- Clone:E5G9V
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Sequence:P41161 T190-Q294
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: ETV5/ERM
Clonality: Monoclonal
Clone: E5G9V
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse