Anti-EED Rabbit Monoclonal Antibody [clone: E4L6E] (Alexa Fluor 647)
Catalog # CSTA98643S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:EED
- Clonality:Monoclonal
- Clone:E4L6E
- Conjugation:Alexa Fluor® 647
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Sequence:CKKQKLSSDENSNPDLSGDENDDAVS
- Storage temperature:4 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: EED
Clonality: Monoclonal
Clone: E4L6E
Conjugation: Alexa Fluor 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey