Anti-Rabex5 Rabbit Monoclonal Antibody [clone: D21F12]
CSTA7622S
- Antibody type:Primary
- Antigen symbol:Rabex5
- Clonality:Monoclonal
- Clone:D21F12
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Sequence:CLEIKPPNQPLAAIDSENVENDK
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Pk:100 µl
Type: Primary
Antigen: Rabex5
Clonality: Monoclonal
Clone: D21F12
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat