Anti-SET1B Rabbit Monoclonal Antibody [clone: D1U5D]
Catalog # CSTA44922S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:SET1B
- Clonality:Monoclonal
- Clone:D1U5D
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Mouse
- Western blot:Yes
- Sequence:CQTLPVGELDAVSPIVNET
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: SET1B
Clonality: Monoclonal
Clone: D1U5D
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Monkey