Anti-SUZ12 Rabbit Monoclonal Antibody [clone: D39F6]
Supplier: Cell Signaling
Certificates
About this item
Type: Primary
Antigen: SUZ12
Clonality: Monoclonal
Clone: D39F6
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey
Specifications
- Catalog No:
- CSTA3737T
- CSTA3737S
- Antigen symbol:
- SUZ12
- SUZ12
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- D39F6
- D39F6
- Reactivity:
- Monkey, Human, Rat, Mouse
- Monkey, Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- ChIP:
- Yes
- Yes
- Sequence:
- CRVTRPGRREFNGMINGETNE
- CRVTRPGRREFNGMINGETNE
- Storage temperature:
- −20 °C
- −20 °C
- Shipping temperature:
- Ambient
- Ambient
- Tested applications:
- CUT&RUN
- CUT&RUN
Specifications
Frequently Bought Together