Anti-TCF7 Rabbit Monoclonal Antibody [clone: C63D9]
Supplier: Cell Signaling
Certificates
About this item
Type: Primary
Antigen: TCF7
Clonality: Monoclonal
Clone: C63D9
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- CSTA2203S
- CSTA2203T
- Antigen symbol:
- TCF7
- TCF7
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- C63D9
- C63D9
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- ChIP:
- Yes
- Yes
- Sequence:
- CGREHRAQRLFPDKLPEPLEDGLKAP
- CGREHRAQRLFPDKLPEPLEDGLKAP
- Storage temperature:
- −20 °C
- −20 °C
- Shipping temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together